Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LIN52 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA564882
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA564882 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA564882 Supplier Invitrogen™ Supplier No. PA564882
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GWKMASPTDGT DLEASLLSFE KLDRASPDLW PEQLPGVAEF AASFKSPITS SPPKWMAEIE RDDIDMLKEL GSLTTANLME KVRGLQNLAY QLGLDESREM TRGKFLNILE KP Highest antigen sequence identity to the following orthologs - mouse 100%, rat 85%.

LIN52 is a protein coding gene. Gene Ontology (GO) annotations related to this gene include DRM complex; transcription, DNA-templated.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LIN52
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.20 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene LIN52
Gene Accession No. Q52LA3
Gene Alias c14_5549; C14orf46; LIN52; lin-52 DREAM MuvB core complex component; lin-52 homolog; Protein lin-52 homolog
Gene Symbols LIN52
Host Species Rabbit
Immunogen Recombinant Human LIN52. Recombinant protein control fragment (Product #RP-88862).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 91750
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.