Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LIN54 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA563326
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA563326 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA563326 Supplier Invitrogen™ Supplier No. PA563326
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VLALPHSQLP QAQKVTTQAQ SGDAKLPPQQ IKVVTIGGRP EVKPVIGVSA LTPGSQLINT TTQPSVLQTQ QLK Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 96%.

LIN54 is a component of the LIN, or DREAM, complex, an essential regulator of cell cycle genes (Schmit et al., 2009 [PubMed 19725879]).
TRUSTED_SUSTAINABILITY

Specifications

Antigen LIN54
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene LIN54
Gene Accession No. Q6MZP7
Gene Alias 4832414L08; AI461788; CXC domain-containing protein 1; CXCDC1; JC8.6; KIAA2037; Lin54; lin-54 DREAM MuvB core complex component; lin-54 homolog; lin-54 homolog (C. elegans); MIP120; mKIAA2037; protein lin-54 homolog; RGD1311361; TCX1
Gene Symbols LIN54
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human LIN54. Recombinant protein control fragment (Product #RP-100284).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 132660
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.