Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LIN7B Antibody (OTI1C9), Allophycocyanin/Cy7, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272131APCCY7
Description
LIN7B Monoclonal specifically detects LIN7B in Human, Mouse, Rat samples. It is validated for Flow Cytometry.Specifications
LIN7B | |
Monoclonal | |
APC-Cyanine7 | |
PBS with 0.05% Sodium Azide | |
LIN7B | |
Full length recombinant protein of human LIN7B (NP_071448) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Flow Cytometry | |
OTI1C9 | |
Flow Cytometry | |
hLin7B, lin-7 homolog B (C. elegans), LIN-7B, MALS-2protein lin-7 homolog B, MALS2veli-2, Mammalian lin-seven protein 2, Veli-2, VELI2hVeli2, Vertebrate lin-7 homolog 2 | |
Mouse | |
Affinity Purified | |
Signal Transduction | |
64130.0 | |
Store at 4°C in the dark. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction