Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LIN7B Monoclonal Antibody (OTI1C9)
GREENER_CHOICE

Catalog No. PIMA525197
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA525197 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA525197 Supplier Invitrogen™ Supplier No. MA525197
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

LIN7B Monoclonal Antibody for Western Blot, ICC/IF, IHC (P), Flow

Plays a role in establishing and maintaining the asymmetric distribution of channels and receptors at the plasma membrane of polarized cells. Forms membrane-associated multiprotein complexes that may regulate delivery and recycling of proteins to the correct membrane domains. The tripartite complex composed of LIN7 (LIN7A, LIN7B or LIN7C), CASK and APBA1 may have the potential to couple synaptic vesicle exocytosis to cell adhesion in brain. Ensures the proper localization of GRIN2B (subunit 2B of the NMDA receptor) to neuronal postsynaptic density and may function in localizing synaptic vesicles at synapses where it is recruited by beta-catenin and cadherin. Required to localize Kir2 channels, GABA transporter (SLC6A12) and EGFR/ERBB1, ERBB2, ERBB3 and ERBB4 to the basolateral membrane of epithelial cells. May increase the amplitude of ACCN3 acid-evoked currents by stabilizing the channel at the cell surface.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LIN7B
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Monoclonal
Clone OTI1C9
Concentration 0.58 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene LIN7B
Gene Accession No. Q9HAP6
Gene Alias hLin7B; hVeli2; lin 7 homolog b; lin-7 homolog B; lin-7 homolog B (C. elegans); lin-7 homolog B, crumbs cell polarity complex component; lin-7-A; LIN7B; lin-7B; MALS2; MALS-2; Mammalian lin-seven protein 2; protein lin-7 homolog B; UNQ3116/PRO10200; Veli1a; Veli2; Veli-2; veli-2 protein; vertebrate homolog of C. elegans Lin-7 type 2; vertebrate lin-7 homolog 2
Gene Symbols LIN7B
Host Species Mouse
Immunogen Full length human recombinant protein of LIN7B produced in HEK293T cell.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64130
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.