Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LIN7C Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA542261
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA542261 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA542261 Supplier Invitrogen™ Supplier No. PA542261
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: MAALGEPVRL ERDICRAIEL LEKLQRSGEV PPQKLQALQR VLQSEFCNAV Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 93%.

Protein assembly at the post synaptic density (PSD) of neuronal synapses is mediated in part by protein interactions with PDZ (PSD-95/discs large/zona occludens-1) motifs. The Mammalian LIN-7 (MALS) proteins are a family of small synaptic proteins that are the mammalian homologs of LIN-7, a C. elegans protein essential for vulval development. MALS proteins are also known as Veli (Vertebrate Lin-7) proteins. MALS-3 mRNA is most abundant in kidney, followed by brain, liver, thymus and heart. MALS-3 is undetectable in spleen. In cultured cortical neurons, MALS proteins are clustered together with PSD-95 and NMDA-type glutamate receptors. MALS proteins are suggested to have roles in regulating the recruitment of neurotransmitter receptors to the PSD and in the assembly of proteins involved in synaptic vesicle exocytosis and synaptic junctions.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LIN7C
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene LIN7C
Gene Accession No. Q9NUP9
Gene Alias 9130007B12Rik; AI303698; AU019331; AW125731; D2Ertd520e; EGK_06400; fb75b09; FLJ11215; lin 7 homolog c; lin-7 homolog C; lin-7 homolog C (C. elegans); lin-7 homolog C, crumbs cell polarity complex component; LIN-7 protein 3; Lin7c; LIN-7C; lin-7-C; lin7c protein; MALS3; MALS-3; Mammalian lin-seven protein 3; mLin7C; neuroepithelial polarity protein; protein lin-7 homolog C; Protein lin-7-like protein C; Unknown (protein for MGC:140723); Veli3; veli-3; veli-3 protein; vertebrate homolog of C. elegans Lin-7 type 3; vertebrate lin-7 homolog 3; wu:fb75b09; zgc:101881
Gene Symbols LIN7C
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human LIN7C (aa 1-50).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55327
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.