Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LIN7C Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$181.90 - $487.50
Specifications
Antigen | LIN7C |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Description
LIN7C Polyclonal specifically detects LIN7C in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
LIN7C | |
Polyclonal | |
Purified | |
RUO | |
PBS with 50% glycerol, pH7.3. | |
55327 | |
IgG | |
Affinity purified |
Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
FLJ11215, lin-7 homolog C (C. elegans), Lin-7C, LIN-7-C, MALS-3LIN-7 protein 3, MALS3protein lin-7 homolog C, Mammalian lin-seven protein 3, veli-3, VELI3Lin7c, Vertebrate lin-7 homolog 3 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-70 of human LIN7C (NP_060832.1). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title