Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LIN9 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA556412
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA556412 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA556412 Supplier Invitrogen™ Supplier No. PA556412
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LNKDLNKVLH KVQQYCYELA PDQGLQPADQ PTDMRRRCEE EAQEIVRHAN SSTGQPCVEN ENLTDLISRL TAILLQIKCL A Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 96%.

LIN9 belongs to the lin-9 family and acts as a tumor suppressor. It inhibits DNA synthesis. Its ability to inhibit oncogenic transformation is mediated through its association with RB1. LIN9 plays a role in the expression of genes required for the G1/S transition.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LIN9
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene LIN9
Gene Accession No. Q5TKA1
Gene Alias 2700022J23Rik; BARA; BARPsv; Beta subunit-associated regulator of apoptosis; HuLin-9; LIN9; Lin-9; lin-9 DREAM MuvB core complex component; lin-9 homolog; lin-9 homolog (C. elegans); mLin-9; pRB-associated protein; protein lin-9 homolog; rb related pathway actor; TGS; TGS1; TGS2; TUDOR gene similar 1 protein; TUDOR gene similar protein; Type I interferon receptor beta chain-associated protein
Gene Symbols LIN9
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human LIN9. Recombinant protein control fragment (Product #RP-94496).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 286826
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.