Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Linalool, 97%

Catalog No. aaa14424ae
missing translation for 'orderingAttributeHoverText'
Quantity:
100 mL
500 mL
2500 mL

Catalog No. AAA14424AE

Supplier: Thermo Scientific Chemicals A14424AE

Only null left
Add to cart

CAS: 78-70-6 | C10H18O | 154.253 g/mol

Linalool is used as a scent in 60-80% of perfumed hygiene products and cleaning agents including soaps, detergents, shampoos, and lotions. as a scent in 60-80% of perfumed hygiene products and cleaning agents including soaps, detergents, shampoos, and lotions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Linalool is used as a scent in 60-80% of perfumed hygiene products and cleaning agents including soaps, detergents, shampoos, and lotions. as a scent in 60-80% of perfumed hygiene products and cleaning agents including soaps, detergents, shampoos, and lotions.

Solubility
Soluble in water (1.589 g/L at 25°C).

Notes
Store at 4°C. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 78-70-6
Molecular Formula C10H18O
Molecular Weight (g/mol) 154.253
MDL Number MFCD00008906
InChI Key CDOSHBSSFJOMGT-UHFFFAOYSA-N
Synonym linalool, linalol, 3,7-dimethyl-1,6-octadien-3-ol, allo-ocimenol, beta-linalool, linalyl alcohol, 1,6-octadien-3-ol, 3,7-dimethyl, +--linalool, p-linalool, linanool
PubChem CID 6549
ChEBI CHEBI:17580
IUPAC Name 3,7-dimethylocta-1,6-dien-3-ol
SMILES CC(=CCCC(C)(C=C)O)C

Specifications

Density 0.865
Boiling Point 199°C to 200°C
Flash Point 76°C (168°F)
Assay Percent Range 97%
Refractive Index 1.462
Quantity 100 mL
Beilstein 1721488
Merck Index 14,5495
Solubility Information Soluble in water (1.589g/L at 25°C).
Formula Weight 154.25
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Linalool
Show More Show Less
Hazard Category H227-H315-H317-H319
Hazard Statement GHS H Statement
H315-H319-H335-H227
Precautionary Statement P210-P261-P264b-P272-P280-P302+P352-P305+P351+P338-P333+P313-P362-P370+P378q-P501c
EINECSNumber 201-134-4
RTECSNumber RG5775000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.