Learn More
Linalool, 97%
Description
Linalool is used as a scent in 60-80% of perfumed hygiene products and cleaning agents including soaps, detergents, shampoos, and lotions. as a scent in 60-80% of perfumed hygiene products and cleaning agents including soaps, detergents, shampoos, and lotions.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Linalool is used as a scent in 60-80% of perfumed hygiene products and cleaning agents including soaps, detergents, shampoos, and lotions. as a scent in 60-80% of perfumed hygiene products and cleaning agents including soaps, detergents, shampoos, and lotions.
Solubility
Soluble in water (1.589 g/L at 25°C).
Notes
Store at 4°C. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
Specifications
Specifications
| Density | 0.865 |
| Boiling Point | 199°C to 200°C |
| Flash Point | 76°C (168°F) |
| Assay Percent Range | 97% |
| Refractive Index | 1.462 |
| Quantity | 100 mL |
| Beilstein | 1721488 |
| Merck Index | 14,5495 |
| Solubility Information | Soluble in water (1.589g/L at 25°C). |
| Formula Weight | 154.25 |
| Show More |
Safety and Handling
| Hazard Category | H227-H315-H317-H319 |
| Hazard Statement | GHS H Statement H315-H319-H335-H227 |
| Precautionary Statement | P210-P261-P264b-P272-P280-P302+P352-P305+P351+P338-P333+P313-P362-P370+P378q-P501c |
| EINECSNumber | 201-134-4 |
| RTECSNumber | RG5775000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.