Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LINC00493 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA567009
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA567009 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA567009 Supplier Invitrogen™ Supplier No. PA567009
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SLLYYSRTMAK SSVDQKDGSA SEVPSELSER PKGFYVETVV TYKEDFVPNT EKILNYWKSW TGGPGTEP Highest antigen sequence identity to the following orthologs - mouse 38%, rat 37%.

SMIM26 is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LINC00493
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SMIM26
Gene Accession No. A0A096LP01
Gene Alias LINC00493; long intergenic non-protein coding RNA 493
Gene Symbols SMIM26
Host Species Rabbit
Immunogen Recombinant Human LINC00493. Recombinant protein control fragment (Product #RP-107649).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 388789
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.