Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LINC00521 Antibody, Novus Biologicals™
SDP

Catalog No. NBP179594 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP179594 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP179594 Supplier Novus Biologicals Supplier No. NBP179594
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

LINC00521 Polyclonal specifically detects LINC00521 in Human samples. It is validated for Western Blot.

Specifications

Antigen LINC00521
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1.0 ug/ml
Formulation PBS, 2% Sucrose with 0.09% Sodium Azide
Gene Alias c14_5713, C14orf48, chromosome 14 open reading frame 48, DKFZp434O1910, MGC33356
Gene Symbols LINC00521
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N terminal of human C14orf48The immunogen for this antibody is C14orf48. Peptide sequence MDTGQRADPSNPGDKEGDLQGLWQELYQLQAKQKKLKREVEKHKLFEDYL.
Molecular Weight of Antigen 16 kDa
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 256369
Reconstitution Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer.
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.