Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LINC02145 Monoclonal Antibody (OTI9E9), TrueMAB™, OriGene
SDP

Catalog No. 502544010
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-4010 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-254-4010 Supplier OriGene Supplier No. CF810646
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

LINC02145 Monoclonal antibody specifically detects LINC02145 in Human samples. It is validated for Western Blot

Specifications

Antigen LINC02145
Applications Western Blot
Classification Monoclonal
Clone OTI9E9
Concentration See Label
Conjugate Unconjugated
Formulation PBS with 8% trehalose and no preservative; pH 7.3
Gene OIP5
Gene Accession No. O43482
Gene Alias FLJ33360; FLJ33360 protein; LINC02145
Host Species Mouse
Immunogen Full length human recombinant protein of human OIP5 produced in E.coli.
Purification Method Protein A/G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11339
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Lyophilized
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.