Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ LINE-1 ORF1p Mouse anti-Human, Clone: 4H1

Catalog No. MABC1152MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABC1152MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABC1152MI Supplier MilliporeSigma™ Supplier No. MABC1152
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Detect LINE-1 retrotransposable element ORF1 protein using this mouse monoclonal Anti-LINE-1 ORF1p Anibody, clone 4H1 Immunohistochemistry (Paraffin), Immunoprecipitation, and Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LINE-1 ORF1p
Applications Western Blot, Immunocytochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 4H1
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Gene Symbols LIRE1,LRE1
Host Species Mouse
Immunogen A linear peptide corresponding to 10 amino aicds from the N-terminal region of human LINE-1 ORF1 protein.
Purification Method Protein G Purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Apoptosis and Cancer
Primary or Secondary Primary
Test Specificity Clone 4H1 specifically detects LINE-1 ORF1 protein in MCF7 cells. This clone targets the sequence corresponding to 10 amino acids from the N-terminal region of LINE-1 ORF1p.
Target Species Human
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.