Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Linear alkylbenzenesulfonic acid, 97%

Catalog No. p-7049882
Click to view available options
Quantity:
250 g
1 kg
3 x 1 kg

CAS: 68584-22-5 | C18H30O3S | 326.495 g/mol

Dodecyl Benzene Sulfonic Acid can be used in all types of household, industrial and institutional cleaners including laundry, dishwash, carwash, hard surface and general purpose detergents. It can also be utilized as an intermediate in preparation of emulsifiers and wetting and foaming agents. Unneutralized compound is an effective surfactant in acidic products such as liquid bowl, aluminum and dairy cleaners. It is an important raw material and intermediate used in Organic Synthesis, Pharma synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 68584-22-5
Molecular Formula C18H30O3S
Molecular Weight (g/mol) 326.495
MDL Number MFCD00147445
InChI Key QJRVOJKLQNSNDB-UHFFFAOYSA-N
Synonym p-3-dodecyl benzenesulphonic acid, c10-c16 alkylbenzenesulfonic acid, 3-4-sulfophenyl dodecane, benzenesulfonic acid, 4-1-ethyldecyl, benzenesulfonic acid, c10-16-alkyl derivs., dsstox_cid_7923, 4-dodecan-3-yl benzenesulfonic acid, c10-c16 alkyl benzene sulfonic acid
PubChem CID 29249
IUPAC Name 4-dodecan-3-ylbenzenesulfonic acid
SMILES CCCCCCCCCC(CC)C1=CC=C(C=C1)S(=O)(=O)O

Specifications

Quantity 250 g
UN Number UN2586
Solubility Information It is soluble in water. 300,000mg/L at 20°C .
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Linear alkylbenzenesulfonic acid
Hazard Category H302-H314-H335
Hazard Statement GHS H Statement
H314-H318-H302-H312
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Precautionary Statement P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: ARYL SULFONIC ACIDS, LIQUID
EINECSNumber 271-528-9
RTECSNumber DB4290000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.