Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AUDIT™ MicroControls™ Linearity FLQ Special Diabetes for Siemens Atellica/Centaur
SDP

Catalog No. 22158604
Click to view available options
Type:
Linearity
This item is not returnable. View return policy
This item is not returnable. View return policy

Linearity FLQ Special Diabetes Siemens Atellica/Centaur is an IVD device consisting of five levels.

The AUDIT™ MicroControls™ Linearity FLQ Special Diabetes Siemens Atellica/Centaur is intended to simulate human patient samples for use in determining linearity, calibration verification, and the verification of reportable range for the following analytes: C-peptide, Insulin.

Specifications

Content And Storage -15°C or colder (Frozen); 2°C to 8°C (Thawed)
Detectable Analytes C-peptide, Insulin
For Use With (Equipment) Analyzers
Sample Type Whole Blood, Serum, Plasma
Type Linearity
For Use With (Application) To simulate human patient samples for use in determining linearity, calibration verification, and the verification of reportable range for the following analytes: C-peptide, Insulin
Quantity 5 x 2 mL
Stability 2 to 8°C (Thawed)
Form Frozen Liquid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.