Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Linezolid
SDP

Catalog No. 376510 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
37-651-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 37-651-0 Supplier R&D Systems™ Supplier No. 3765/10

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Antibiotic; inhibits protein synthesis in gram-positive bacteria

Oxazolidinone antibiotic. Inhibits bacterial protein synthesis prior to chain initiation. Displays potent antibacterial activity against a variety of multidrug-resistant gram-positive microbes in vitro and in vivo. Identified as targeting human host cell proteins that interact with SARS-CoV-2.

Chemical Identifiers

CAS 165800-03-3
Molecular Formula C16H20FN3O4

Specifications

Quantity 10 mg
Product Line Tocris
Formula Weight 337.35
Purity 0.98
Molecular Weight (g/mol) 337.351
Chemical Name or Material Linezolid
Recommended Storage Store at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.