Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LINGO-1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $610.00
Specifications
Antigen | LINGO-1 |
---|---|
Applications | Immunocytochemistry, Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
LINGO-1 Polyclonal specifically detects LINGO-1 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
LINGO-1 | |
Polyclonal | |
Rabbit | |
Neuroscience | |
FLJ14594, LERN1UNQ201, leucine rich repeat and Ig domain containing 1, leucine rich repeat neuronal 6A, Leucine-rich repeat and immunoglobilin domain-containing protein 1, Leucine-rich repeat neuronal protein 1, Leucine-rich repeat neuronal protein 6A, LRRN6Aleucine-rich repeat neuronal 6A, MGC17422 | |
LINGO1 | |
IgG | |
Affinity Purified |
Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
RUO | |
Human | |
84894 | |
This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:TETRLLDLGKNRIKTLNQDEFASFPHLEELELNENIVSAV | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title