Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LINGO-2 Goat anti-Human, Polyclonal, R&D Systems™

Catalog No. AF3679
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF3679 100 μg
AF3679SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. AF3679 Supplier R&D Systems Supplier No. AF3679
This item is not returnable. View return policy

Goat Polyclonal Antibody

LINGO-2 Polyclonal antibody AF3679 Specifically detects LINGO-2 in Human samples. It is validated for Immunocytochemistry, Western Blotting

Specifications

Antigen LINGO-2
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.1 ug/mL, Immunocytochemistry 5-15 ug/mL
Gene Accession No. Q7L985
Gene Alias LERN3, LERN3FLJ31810, leucine rich repeat and Ig domain containing 2, leucine-rich repeat and immunoglobulin-like domain-containing nogoreceptor-interacting protein 2, Leucine-rich repeat neuronal protein 3, Leucine-rich repeat neuronal protein 6C, LINGO2, LRRN6C, LRRN6Cleucine rich repeat neuronal 6C
Gene Symbols LINGO2
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant human LINGO-2 Cys28-Leu542 Accession # Q7L985
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 158038
Test Specificity Detects human LINGO-2 in direct ELISAs and Western blots. In direct ELISAs, less than 5% cross-reactivity with recombinant human LINGO-1 is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.