Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LINGO-2 Mouse anti-Human, Clone: 382008, R&D Systems™

Catalog No. MAB3679
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB3679 100 μg
MAB3679SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB3679 Supplier R&D Systems Supplier No. MAB3679
This item is not returnable. View return policy

Mouse Monoclonal Antibody

LINGO-2 Monoclonal antibody MAB3679 Specifically detects LINGO-2 in Human samples. It is validated for Western Blotting

Specifications

Antigen LINGO-2
Applications Western Blot
Classification Monoclonal
Clone 382008
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q7L985
Gene Alias LERN3, LERN3FLJ31810, leucine rich repeat and Ig domain containing 2, leucine-rich repeat and immunoglobulin-like domain-containing nogoreceptor-interacting protein 2, Leucine-rich repeat neuronal protein 3, Leucine-rich repeat neuronal protein 6C, LINGO2, LRRN6C, LRRN6Cleucine rich repeat neuronal 6C
Gene Symbols LINGO2
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human LINGO-2 Cys28-Leu542 Accession # Q7L985
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 158038
Test Specificity Detects human LINGO-2 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human LINGO-1 is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.