Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LINGO1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA532898
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA532898 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA532898 Supplier Invitrogen™ Supplier No. PA532898
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Percent identity with other species by BLAST analysis: Human, Gorilla, Orangutan, Monkey, Marmoset, Mouse, Rat, Bovine, Dog, Bat, Elephant, Panda, Horse, Rabbit, Pig (100%). For IHC(P), use heat induced antigen retrieval in pH 6.0 citrate buffer. After incubation with the primary antibody, slides were incubated with biotinylated secondary antibody, followed by alkaline phosphatase-streptavidin and chromogen.

Functional component of the Nogo receptor signaling complex (RTN4R/NGFR) in RhoA activation responsible for some inhibition of axonal regeneration by myelin-associated factors (PubMed:14966521, PubMed:15694321). Is also an important negative regulator of oligodentrocyte differentiation and axonal myelination (PubMed:15895088). Acts in conjunction with RTN4 and RTN4R in regulating neuronal precursor cell motility during cortical development. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen LINGO1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide
Gene LINGO1
Gene Accession No. Q96FE5, Q9D1T0
Gene Alias 4930471K13Rik; AV148400; LERN1; leucine rich repeat and Ig domain containing 1; leucine rich repeat neuronal 6A; leucine-rich repeat; leucine-rich repeat and immunoglobulin domain-containing protein 1; Leucine-rich repeat and immunoglobulin-like domain-containing nogo receptor-interacting protein 1; leucine-rich repeat neuronal 6A; leucine-rich repeat neuronal protein 1; Leucine-rich repeat neuronal protein 6A; Lingo1; LINGO-1; LRRN6A; UNQ201; UNQ201/PRO227
Gene Symbols LINGO1
Host Species Rabbit
Immunogen Synthetic 14 amino acid peptide from C-terminus of human LINGO1.
Purification Method Antigen affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100061441, 100155465, 100344366, 235402, 315691, 487660, 520694, 84894
Target Species Human, Mouse, Rat, Canine, Rabbit, Monkey, Bovine, Pig, Horse, Bat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.