Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LINGO1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA597210
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA597210 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA597210 Supplier Invitrogen™ Supplier No. PA597210
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: CPPRCECSAQ DRAVLCHRKR FVAVPEGIPT ETRLLDLGKN RIKTLNQDEF ASFPHLEELE LNENIVSAVE PGAFNNLFNL RTLGLRSNRL KLIPLGVFTG LSNLTKLDIS ENKIVILLDY MFQDLYNLKS LEVGDNDLVY ISHRAFSGLN SLEQLTLEKC NLTSIPTEAL SHLHGLIVLR LRHLNINAIR DYSFKRLYR; Positive Samples: MCF-7, A-549, K-562, Mouse testis, Mouse lung; Cellular Location: Cell membrane, Single-pass type I membrane protein.

Functional component of the Nogo receptor signaling complex (RTN4R/NGFR) in RhoA activation responsible for some inhibition of axonal regeneration by myelin-associated factors (PubMed:14966521, PubMed:15694321). Is also an important negative regulator of oligodentrocyte differentiation and axonal myelination (PubMed:15895088). Acts in conjunction with RTN4 and RTN4R in regulating neuronal precursor cell motility during cortical development. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen LINGO1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.13 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene LINGO1
Gene Accession No. Q96FE5, Q9D1T0
Gene Alias 4930471K13Rik; AV148400; LERN1; leucine rich repeat and Ig domain containing 1; leucine rich repeat neuronal 6A; leucine-rich repeat; leucine-rich repeat and immunoglobulin domain-containing protein 1; Leucine-rich repeat and immunoglobulin-like domain-containing nogo receptor-interacting protein 1; leucine-rich repeat neuronal 6A; leucine-rich repeat neuronal protein 1; Leucine-rich repeat neuronal protein 6A; Lingo1; LINGO-1; LRRN6A; UNQ201; UNQ201/PRO227
Gene Symbols LINGO1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 42-240 of human LINGO1 (NP_1161974).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 235402, 84894
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.