Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LINGO2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA599869
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA599869 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA599869 Supplier Invitrogen™ Supplier No. PA599869
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total LINGO2.

Leucine-rich repeat and immunoglobulin-like domain-containing protein 2 (LINGO2) is a gene located on chromosome 9q34.3, encoding a transmembrane protein predominantly expressed in the central nervous system. Structurally, LINGO2 features leucine-rich repeat (LRR) domains and immunoglobulin-like domains, which are crucial for molecular interactions and signaling. Functionally, LINGO2 is involved in regulating neural growth and development, influencing processes such as axon guidance and synaptic plasticity. It modulates neurotransmitter receptor activity and intracellular signaling pathways essential for neural transmission, with implications for both normal cognitive function and neurological disorders. Altered expression of LINGO2 has been associated with movement disorders, as well as neurodegenerative conditions, suggesting its role in neural circuit integrity. Current research on LINGO2 is exploring its potential as a therapeutic target for enhancing neural repair mechanisms and addressing synaptic dysfunction, providing insights into interventions for neurological diseases.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LINGO2
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene LINGO2
Gene Accession No. Q3URE9, Q7L985
Gene Alias B230217C06Rik; LERN3; LERN3FLJ31810; leucine rich repeat and Ig domain containing 2; leucine rich repeat neuronal 6C; leucine-rich repeat and immunoglobulin-like domain-containing nogo receptor-interacting protein 2; Leucine-rich repeat neuronal protein 3; leucine-rich repeat neuronal protein 6C; Lingo2; LRRN6C; RGD1310773; UNQ9234/PRO31993
Gene Symbols LINGO2
Host Species Rabbit
Immunogen A synthesized peptide derived from human LINGO2(Accession Q7L985), corresponding to amino acid residues L364-E414.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 158038, 242384, 313156
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.