Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LINGO3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565368
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565368 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565368 Supplier Invitrogen™ Supplier No. PA565368
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LWIVQRRKTLN FDGRLPACAT PAEVRGDALR NLPDSVLFEY FVCRKPKIRE RRLQRVTATA GEDVRFLCR Highest antigen sequence identity to the following orthologs - mouse 94%, rat 94%.

LINGO3 is a protein coding gene. Gene Ontology (GO) annotations related to this gene include extracellular matrix; extracellular space; integral component of membrane.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LINGO3
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene LINGO3
Gene Accession No. P0C6S8
Gene Alias hCG2040376; LERN2; leucine rich repeat and Ig domain containing 3; leucine rich repeat neuronal 6B; leucine-rich repeat and immunoglobulin-like domain-containing nogo receptor-interacting protein 3; Leucine-rich repeat neuronal protein 2; leucine-rich repeat neuronal protein 6B; LINGO3; LRRN6B
Gene Symbols LINGO3
Host Species Rabbit
Immunogen Recombinant Human LINGO3. Recombinant protein control fragment (Product #RP-100012).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 645191
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.