Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LINGO4 Rabbit anti-Human, Polyclonal, Bioss

Rabbit Polyclonal Antibody
Supplier: Bioss BS11873R

Description
LINGO4 is a protein coding gene.Specifications
LINGO4 | |
Polyclonal | |
Unconjugated | |
LINGO4 | |
DAAT9248; leucine rich repeat and Ig domain containing 4; Leucine-rich repeat and immunoglobulin-like domain-containing nogo receptor-interacting protein 4; Leucine-rich repeat neuronal protein 6D; LINGO4; LRRN6D; PRO34002; UNQ9248/PRO34002 | |
Rabbit | |
Protein A | |
RUO | |
339398 | |
-20°C | |
Liquid |
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
1 μg/mL | |
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH | |
Q6UY18 | |
LINGO4 | |
KLH conjugated synthetic peptide derived from human LINGO4. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction