Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Linoleic Acid Methyl Ester, MP Biomedicals™

Catalog No. ICN10080705 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
25 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10080705 5 g
ICN10080701 1 g
ICN10080710 10 g
ICN10080725 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. ICN10080705 Supplier MP Biomedicals, Inc Supplier No. 0210080705
Add to Cart
Add to Cart
Molecular biology
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 112-63-0
Molecular Formula C19H34O2
Molecular Weight (g/mol) 294.479
MDL Number MFCD00009534
InChI Key WTTJVINHCBCLGX-NQLNTKRDSA-N
Synonym methyl linoleate, linoleic acid methyl ester, methyl lineoleate, methyl 9z,12z-octadeca-9,12-dienoate, methyl octadecadienoate, methyl 9-cis,12-cis-octadecadienoate, linoleic acid,methyl ester, linoleic acid, methyl ester, methyl linoleate, native, 9,12-octadecadienoic acid z,z-, methyl ester
PubChem CID 5284421
ChEBI CHEBI:69080
IUPAC Name methyl (9Z,12Z)-octadeca-9,12-dienoate
SMILES CCCCCC=CCC=CCCCCCCCC(=O)OC

Specifications

Color Colorless
Density 0.889g/mL
Quantity 5 g
Formula Weight 294.5
Physical Form Liquid
Chemical Name or Material Linoleic Acid Methyl Ester
Recommended Storage Store at 0°C.
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.