Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Linoleic acid, tech. 75%

Catalog No. aaa1526509
Click to view available options
Quantity:
10 g
50 g
250 g

CAS: 60-33-3 | C18H32O2 | 280.452 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 60-33-3
Molecular Formula C18H32O2
Molecular Weight (g/mol) 280.452
MDL Number MFCD00064241
InChI Key OYHQOLUKZRVURQ-HZJYTTRNSA-N
Synonym linoleic acid, linolic acid, telfairic acid, cis,cis-linoleic acid, 9z,12z-octadeca-9,12-dienoic acid, linoleate, grape seed oil, cis,cis-9,12-octadecadienoic acid, cis-9,cis-12-octadecadienoic acid, 9z,12z-linoleic acid
PubChem CID 5280450
ChEBI CHEBI:17351
IUPAC Name (9Z,12Z)-octadeca-9,12-dienoic acid
SMILES CCCCCC=CCC=CCCCCCCCC(=O)O

Specifications

Density 0.902
Boiling Point 230°C (16 mm)
Flash Point >110°C (230°F)
Assay Percent Range 75%
Refractive Index 1.4699
Linear Formula CH3(CH2)4CH=CHCH2CH=CH(CH2)7CO2H
Quantity 10 g
Beilstein 1727101
Sensitivity Air sensitive
Merck Index 14,5505
Formula Weight 280.45
Percent Purity 75%
Grade Technical
Physical Form Liquid
Chemical Name or Material Linoleic Acid
Show More Show Less
EINECSNumber 200-470-9
RTECSNumber RF9990000
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.