Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Linolenic acid, 55%, MP Biomedicals™

Catalog No. ICN10218890 Shop All MP Biomedicals, Inc Products
Click to view available options
Quantity:
100 g
500 g
1 kg

Chemical Identifiers

CAS 463-40-1
Molecular Formula C18H30O2
Molecular Weight (g/mol) 278.436
InChI Key DTOSIQBPPRVQHS-PDBXOOCHSA-N
Synonym linolenic acid, alpha-linolenic acid, linolenate, 9z,12z,15z-octadeca-9,12,15-trienoic acid, a-linolenic acid, cis,cis,cis-9,12,15-octadecatrienoic acid, all-cis-9,12,15-octadecatrienoic acid, alpha-linolenate, 9-cis,12-cis,15-cis-octadecatrienoic acid, z,z,z-9,12,15-octadecatrienoic acid
PubChem CID 5280934
ChEBI CHEBI:27432
IUPAC Name (9Z,12Z,15Z)-octadeca-9,12,15-trienoic acid
SMILES CCC=CCC=CCC=CCCCCCCCC(=O)O

Specifications

Color Colorless
Density 0.916 at 20°C (water = 1)
Quantity 500 g
Formula Weight 278.4
Percent Purity 0.55
Physical Form Liquid
Chemical Name or Material Linolenic acid
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.