Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Linopirdine dihydrochloride
GREENER_CHOICESDP

Catalog No. 199910 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
19-991-0 10 mg
19-995-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-991-0 Supplier Tocris Bioscience™ Supplier No. 1999/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

KV7 (KCNQ) channel blocker

Blocker of KV7 (KCNQ) voltage-gated potassium channels; blocks KV7.1 + 7.3 (KCNQ2 + 3)/ M-currents (IC50 = 4 - 7 μM) and KV7.1 (KCNQ1) homomeric channels (IC50 = 8.9 μM). Augments hippocampal ACh release and is a cognitive enhancer following oral administration in vivo.

Chemical Identifiers

CAS 113168-57-3
Molecular Formula C26H23Cl2N3O

Specifications

Quantity 10 mg
Formula Weight 464.39
Purity >99%
Molecular Weight (g/mol) 464.39
Chemical Name or Material Linopirdine dihydrochloride
Recommended Storage Desiccate at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.