Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ LINS Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA565215
Description
Immunogen sequence: TESKYDISICG CVPSLVQDQS SNQTIPHRLT APHSHRDVCA RHSWASDAPS EPLKAVMSKG AHTMCASSLS S Highest antigen sequence identity to the following orthologs - mouse 28%, rat 25%.
The Drosophila segment polarity gene lin encodes a protein, lines, which plays important roles in development of the epidermis and hindgut. This gene encodes a protein containing a lines-like domain. This gene is located on chromosome 15 and clustered with the gene encoding ankyrin repeat and SOCS box-containing protein 7. Alternative splicing results in multiple transcript variants.
Specifications
| LINS | |
| Polyclonal | |
| Unconjugated | |
| LINS1 | |
| lines homolog 1; LINS; LINS1; MRT27; Protein Lines homolog 1; WINS1; WINS1 protein with Drosophila Lines (Lin) homologous domain; wnt-signaling molecule Lines homolog 1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 55180 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8NG48 | |
| LINS1 | |
| Recombinant Human LINS. Recombinant protein control fragment (Product #RP-105953). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction