Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LINS Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA558450
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA558450 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA558450 Supplier Invitrogen™ Supplier No. PA558450
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SRVFIEQDDD MLEAAKASLG IYLTLTRGCE ATESLTQGKE MWDHHTHENG YNPHCIFLFF LKNIGFDSTV LLDFLISSET CFLEYFVRYL KLLQKDWDN Highest antigen sequence identity to the following orthologs: Mouse - 79%, Rat - 81%.

The Drosophila segment polarity gene lin encodes a protein, lines, which plays important roles in development of the epidermis and hindgut. This gene encodes a protein containing a lines-like domain. This gene is located on chromosome 15 and clustered with the gene encoding ankyrin repeat and SOCS box-containing protein 7. Alternative splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LINS
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene LINS1
Gene Accession No. Q8NG48
Gene Alias lines homolog 1; LINS; LINS1; MRT27; Protein Lines homolog 1; WINS1; WINS1 protein with Drosophila Lines (Lin) homologous domain; wnt-signaling molecule Lines homolog 1
Gene Symbols LINS1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human LINS. Recombinant protein control fragment (Product #RP-98107).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55180
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.