Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Poltex Conical Tube Rack Holder
SDP

Catalog No. 22022581
Click to view available options
Mounting:
Magnet
Tape
Wall

Poltex conical tube rack holder is a convenient holder for the Conical Tube Holder (CONIC) line of racks.

  • Convenient holder for the Conical Tube Holder (CONIC) line of racks
  • Holds up to 5 CONIC racks
  • Durable and cleanable PETG construction
  • Magnet mount (powerful magnets included), Tape mount (VHB Tape Included), or Wall mount (screws/anchors included)
  • Comes with rubber bumpons on base for optional countertop placement

Specifications

Product Type Tube Rack
Autoclavable Not Autoclavable
Color Clear
Holds Tube Racks
Material Plastic
Positions 1
Shape Rectangular
Shelves None
Stackable No
Sterility Non-sterile
Mounting Wall
Length (Metric) 101.60 mm
Width (Metric) 173.99 mm
Height (Metric) 266.70 mm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.