Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LIPA Monoclonal Antibody (9G7F12)
GREENER_CHOICE

Catalog No. PIMA515278
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA515278 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA515278 Supplier Invitrogen™ Supplier No. MA515278
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA5-15278 targets LAL in indirect ELISA, WB applications and shows reactivity with Human samples. The MA5-15278 immunogen is purified recombinant fragment of LAL expressed in E. Coli.

This gene encodes lipase A, the lysosomal acid lipase (also known as cholesterol ester hydrolase). This enzyme functions in the lysosome to catalyze the hydrolysis of cholesteryl esters and triglycerides. Mutations in this gene can result in Wolman disease and cholesteryl ester storage disease. Alternatively spliced transcript variants encoding the same protein have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LIPA
Applications ELISA, Western Blot
Classification Monoclonal
Clone 9G7F12, 7G6D7
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation Ascites with 0.03% sodium azide
Gene LIPA
Gene Accession No. P38571
Gene Alias AA960673; Acid cholesteryl ester hydrolase; CESD; Chole; Chole2; cholesterol ester hydrolase; Cholesterol esterase (pancreatic) see D3Wox12 D3Wox13 D3Wox26 and D3Mgh25; cholesteryl esterase; HGCLAS; HUSSY-01; LAL; LAS; LIAS; Lip1; Lip-1; Lipa; Lipase A; lipase A, lysosomal acid type; lipase A, lysosomal acid, cholesterol esterase; lipoate synthase; Lipoic acid synthase; lipoic acid synthetase; lipoyl synthase, mitochondrial; lip-syn; LS; lysosomal acid lipase; lysosomal acid lipase 1; lysosomal acid lipase A; lysosomal acid lipase/cholesteryl ester hydrolase; pancreatic cholesterol esterase; PDHLD; RP11-341B24.1; Sterol esterase
Gene Symbols LIPA
Host Species Mouse
Immunogen Purified recombinant fragment of LAL expressed in E. Coli.
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3988
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.