Learn More
Invitrogen™ LIPA Polyclonal Antibody

Description
The target sequence has 82% sequence homology with porcine and 73% sequence homology with rat and bovine.
Specifications
Specifications
| Antigen | LIPA |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1.0 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide |
| Gene | LIPA |
| Gene Accession No. | P38571, Q9Z0M5 |
| Gene Alias | AA960673; Acid cholesteryl ester hydrolase; CESD; Chole; Chole2; cholesterol ester hydrolase; Cholesterol esterase (pancreatic) see D3Wox12 D3Wox13 D3Wox26 and D3Mgh25; cholesteryl esterase; HGCLAS; HUSSY-01; LAL; LAS; LIAS; Lip1; Lip-1; Lipa; Lipase A; lipase A, lysosomal acid type; lipase A, lysosomal acid, cholesterol esterase; lipoate synthase; Lipoic acid synthase; lipoic acid synthetase; lipoyl synthase, mitochondrial; lip-syn; LS; lysosomal acid lipase; lysosomal acid lipase 1; lysosomal acid lipase A; lysosomal acid lipase/cholesteryl ester hydrolase; pancreatic cholesterol esterase; PDHLD; RP11-341B24.1; Sterol esterase |
| Gene Symbols | LIPA |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.