Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lipase Monoclonal Antibody (LIP10-106.4), QED Bioscience™
SDP

Catalog No. 502277508
Encompass
Change view
Click to view available options
Quantity:
1 mg
100 μg
500 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-227-7508 500 μg
50-227-7507 100 μg
50-227-7506 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 50-227-7508 Supplier Leinco Technologies Inc Supplier No. 13301500UG
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Lipase Monoclonal antibody specifically detects Lipase in Human samples. It is validated for ELISA

Specifications

Antigen Lipase
Applications ELISA
Classification Monoclonal
Clone LIP10-106.4
Concentration 0.5-1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene PNLIP
Gene Accession No. P16233
Gene Alias 1810007A24Rik; pancreatic lipase; pancreatic triacylglycerol lipase; pancreatic triglyceride lipase; PANLI; PL; PNLIP; PNLIPD; PNLIPRP2; PTL; RP11-33D13.1; triglyceride lipase
Host Species Mouse
Immunogen Purified human pancreatic lipase.
Purification Method Protein A
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5406
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.