Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LIPC Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA578040
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA578040 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA578040 Supplier Invitrogen™ Supplier No. PA578040
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Control: HepG2 Predicted Reactivity: Mouse (81%), Dog (82%), Pig (80%), Rabbit (82%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

The Lipase family belongs to one of the most robust genetic superfamilies found in living organisms that includes esterases and thioesterases. Lipase gene products are related by tertiary structure rather than primary amino acid sequence. Balancing the composition and the transport of lipoproteins in human plasma is essential for normal body function and is mediated in part by Hepatic Lipase, also known as HL or LIPC. Rare deficiencies in Hepatic Lipase have been identified in humans which lead to pathologic levels of circulating lipoprotein particles; this condition is associated with coronary artery disease (CAD). Hepatic Lipase is regulated by thyroid hormones and has a dual function as a triglyceride hydrolase and a ligand/bridging factor for receptor-mediated lipoprotein uptake. Hepatic Lipase localizes to the endothelial surfaces of extrahepatic tissues.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LIPC
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
Gene LIPC
Gene Accession No. P11150
Gene Alias AI256194; HDLCQ12; hepatic lipase; hepatic triacylglycerol lipase; HL; Hpl; HTGL; lipase C; lipase C, hepatic type; lipase member C; lipase, hepatic; LIPC; LIPH; Triacylglycerol lipase
Gene Symbols LIPC
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human Hepatic lipase. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3990
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.