Learn More
Description
Used for: imaging lipid peroxides in living cells. Liperfluo can be used to monitor lipid peroxidation in ferroptosis research.
Application: fluorescent microscopy and flow cytometry.
Properties and Photophysical Data: Liperfluo has a high solubility in organic solvents. It displays minimal photo damage and auto-fluorescence and shows high selectivity to lipid peroxidases over ROS and other radical species. Excitation and emission maxima (λ) measured in ethanol are 524 nm and 535 nm, respectively; quantum yield (oxidized form) = 0.85.
Please refer to this protocol for further information on how to use this product.
Specifications
Specifications
| Chemical Name or Material | 2-[4-(Diphenylphosphino)phenyl]-9-(3,6,9,12-tetraoxatridec-1-yl)anthra[2,1,9-def :6,5,10-d'e'f']diisoquinoline-1,3,8,10(2H ,9H)-tetrone |
| CAS | 1448846-35-2 |
| Quantity | 50 μg |
| Product Line | Tocris |
| Target | Fluorescent Lipids Probes and Cell Membrane Stains |
| Molecular Formula | C51H41N2O8P |
| Purity | 90% |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
