Learn More
Invitrogen™ LIPF Polyclonal Antibody

Description
The target sequence has 87% sequence homology with canine. Suggested positive control: human and mouse liver lysate, antigen standard for LIPG (transient overexpression lysate).
Specifications
Specifications
| Antigen | LIPF |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.9 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 30% glycerol and 0.1% sodium azide |
| Gene | LIPF |
| Gene Accession No. | P07098, Q9CPP7 |
| Gene Alias | 2310051B21Rik; AV082900; DGL; EDL; EL; endothelial lipase; gastric lipase; gastric triacylglycerol lipase; GL; HGL; HLAL; lingual lipase; lipase F; lipase F, gastric type; lipase, gastric; LIPE; LIPF; MGC138477; MGC142271; prelingual lipase; RNLIP; Rnlp; triacylglycerol lipase |
| Gene Symbols | LIPF |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.