Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LIPF Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA596545
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA596545 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA596545 Supplier Invitrogen™ Supplier No. PA596545
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: QSLFKFIFGD KIFYPHNFFD QFLATEVCSR EMLNLLCSNA LFIICGFDSK NFNTSRLDVY LSHNPAGTSV QNMFHWTQAV KSGKFQAYDW GSPVQNRMHY DQSQPPYYNV TAMNVPIAVW NGGKDLLADP QDVGLLLPKL PNLIYHKEIP FYNHLDFIWA MDAPQEVYND IVSMISEDKK; Positive Samples: MCF7, Mouse small intestine, Mouse lung, Mouse heart, Mouse liver, Mouse stomach, Rat kidney.

NELF (nasal embryonic luteinizing hormone-releasing hormone factor) is a 530 amino acid transcription factor involved in the migration of LHRH neurons, outgrowth of olfactory axons and suppression of transcription elongation. NELF is found in the peripheral and central nervous system during embryonic development, and is highly expressed in adult testis, kidney and brain. Known to couple NMDA receptor signaling to the nucleus, NELF knockdown impaired GnRH neuronal migration of NLT cells in vitro and the gene encoding NELF has been linked to the development of Idiopathic hypogonadotropic hypogonadism (IHH), a disorder resulting in impaired pubertal maturation and reproductive function.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LIPF
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 2.41 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene LIPF
Gene Accession No. P04634, P07098, Q9CPP7
Gene Alias 2310051B21Rik; AV082900; DGL; EDL; EL; endothelial lipase; gastric lipase; gastric triacylglycerol lipase; GL; HGL; HLAL; lingual lipase; lipase F; lipase F, gastric type; lipase, gastric; LIPE; LIPF; MGC138477; MGC142271; prelingual lipase; RNLIP; Rnlp; triacylglycerol lipase
Gene Symbols LIPF
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 219-398 of human LIPF (NP_0041811).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 50682, 67717, 8513
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.