Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LIPG Monoclonal Antibody (OTI7C2), TrueMAB™, OriGene
SDP

Catalog No. 502542582
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-2582 30 μL
50-167-2183 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-254-2582 Supplier OriGene Supplier No. TA501040S
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

LIPG Monoclonal antibody specifically detects LIPG in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot

Specifications

Antigen LIPG
Applications Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Monoclonal
Clone OTI7C2
Concentration 0.89 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene LIPG
Gene Accession No. Q9Y5X9
Gene Alias 3110013K01Rik; EDL; EL; endothelial cell-derived lipase; Endothelial lipase; endothelial-derived lipase; lipase; lipase G; lipase G, endothelial type; lipase, endothelial; LIPG; lipoprotein lipase H; lipose, endothelial; mEDL; PL; PRO719; UNQ387/PRO719
Host Species Mouse
Immunogen Full length human recombinant protein of human LIPG produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9388
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.