Learn More
LIPG Mouse anti-Human, Clone: OTI4C3, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF501020

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
LIPG (Lipase G, Endohelial type) is an endothelial lipase which is a novel member of the lipoprotein lipase gene family. LIPG is synthesized by endothelial cells and functions at the site of synthesis. LIPG most likely plays a physiological role in modulating lipoprotein metabolism. Overexpressed LIPG has been shown to reduce HDL cholesterol levels, whereas an LIPG deficiency appears to be effective in elevating HDL cholesterol levels in animals. LIPG has substantial phospholipase activity and may be involved in lipoprotein metabolism and vascular biology. Also, LIPG is designated a member of the TG lipase family by its sequence and characteristic lid region which provides substrate specificity for enzymes of the TG lipase family. LIPG is a carboxyl esterase that hydrolyzes high density lipoproteins, emulsifies triglycerides, is essential for the efficient digestion of dietary fats and can bind to heparin.Specifications
LIPG | |
Monoclonal | |
Unconjugated | |
LIPG | |
EDL, EL, PRO719 | |
Mouse | |
Affinity Chromatography | |
RUO | |
9388 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation | |
OTI4C3 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q9Y5X9 | |
LIPG | |
Full length human recombit protein of human LIPG produced in HEK293T cell. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2b |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.