Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LIPG Polyclonal Antibody, Biotin
GREENER_CHOICE

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA522916

Catalog No. PIPA522916


Only null left
Add to Cart

Description

Description

This antibody does not react with mouse endothelial lipase.

LIPG (Lipase G, Endohelial type) is an endothelial lipase which is a novel member of the lipoprotein lipase gene family. LIPG is synthesized by endothelial cells and functions at the site of synthesis. LIPG most likely plays a physiological role in modulating lipoprotein metabolism. Overexpressed LIPG has been shown to reduce HDL cholesterol levels, whereas an LIPG deficiency appears to be effective in elevating HDL cholesterol levels in animals. LIPG has substantial phospholipase activity and may be involved in lipoprotein metabolism and vascular biology. Also, LIPG is designated a member of the TG lipase family by its sequence and characteristic lid region which provides substrate specificity for enzymes of the TG lipase family. LIPG is a carboxyl esterase that hydrolyzes high density lipoproteins, emulsifies triglycerides, is essential for the efficient digestion of dietary fats and can bind to heparin.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

LIPG
Polyclonal
Biotin
LIPG
3110013K01Rik; EDL; EL; endothelial cell-derived lipase; Endothelial lipase; endothelial-derived lipase; lipase; lipase G; lipase G, endothelial type; lipase, endothelial; LIPG; lipoprotein lipase H; lipose, endothelial; mEDL; PL; PRO719; UNQ387/PRO719
Rabbit
Antigen affinity chromatography
RUO
9388
4°C, store in dark
Liquid
ELISA, Western Blot, Immunocytochemistry
0.9 mg/mL
PBS with 0.05% sodium azide
Q9Y5X9
LIPG
An N-terminal synthetic peptide made to human Endothelial Lipase.
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.