Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LIPH Mouse anti-Human, Porcine, Clone: 1F5D10, Proteintech

Mouse Monoclonal Antibody
$518.60
Specifications
Antigen | LIPH |
---|---|
Clone | 1F5D10 |
Concentration | 2 mg/mL |
Applications | Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-6735
![]() ![]() |
Proteintech Group Inc
663031IG150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
This gene encodes a membrane-bound member of the mammalian triglyceride lipase family. It catalyzes the production of 2-acyl lysophosphatidic acid (LPA), which is a lipid mediator with diverse biological properties that include platelet aggregation, smooth muscle contraction, and stimulation of cell proliferation and motility.Specifications
LIPH | |
2 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
AH, Lipase member H, lipase, member H, LIPH, LPD lipase related protein, LPDLR, mPA PLA1, mPA PLA1 alpha, MPAPLA1, Phospholipase A1 member B, PLA1B | |
LIPH | |
IgG2a | |
Protein A | |
Antibody |
1F5D10 | |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Mouse | |
Human, Pig | |
Q8WWY8 | |
100737989, 200879 | |
LIPH Fusion Protein Ag24009 | |
Primary | |
-20°C | |
LIPH |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title