Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LIPH Polyclonal Antibody

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA5109919

Catalog No. PIPA5109919


Only null left
Add to Cart

Description

Description

Immunogen sequence: KLRDKAGNTT ESKINHEPTT FQKYHQVSLL ARFNQDLDKV AAISLMFSTG SLIGPRYKLR ILRMKLRSLA HPERPQLCRY DLVLMENVET VFQPILCPEL Q.

This gene encodes a membrane-bound member of the mammalian triglyceride lipase family. It catalyzes the production of 2-acyl lysophosphatidic acid (LPA), which is a lipid mediator with diverse biological properties that include platelet aggregation, smooth muscle contraction, and stimulation of cell proliferation and motility.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

LIPH
Polyclonal
Unconjugated
LIPH
AH; ARWH2; C130037N08Rik; D16Wsu119e; H06; HYPT7; Lacrimal lipase; LAH2; lipase H; lipase member H; lipase, member H; LIPH; LPD lipase-related protein; lpd1; lpd2; LPDLR; Membrane-associated phosphatidic acid-selective phospholipase A1-alpha; membrane-bound phosphatidic acid-selective phospholipase A1; MPAPLA1; mPA-PLA1; mPA-PLA1 alpha; P3; phospholipase A(1); phospholipase A1 member B; PLA1B
Rabbit
Affinity Chromatography
RUO
200879, 239759, 681694
-20°C, Avoid Freeze/Thaw Cycles
Liquid
ELISA, Western Blot
1.53 mg/mL
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Q32PY2, Q8CIV3, Q8WWY8
LIPH
A synthetic peptide corresponding to a sequence within amino acids 350-450 of human LIPH (NP_640341.1).
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.