Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LIPI Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA549416

Catalog No. PA549416


Only null left
Add to Cart

Description

Description

LIPI Polyclonal Antibody for Western Blot

Hydrolyzes specifically phosphatidic acid (PA) to produce 2-acyl lysophosphatidic acid (LPA; a potent bioactive lipid mediator) and fatty acid. Does not hydrolyze other phospholipids, like phosphatidylserine (PS), phosphatidylcholine (PC) and phosphatidylethanolamine (PE) or triacylglycerol (TG).
TRUSTED_SUSTAINABILITY
Specifications

Specifications

LIPI
Polyclonal
Unconjugated
LIPI
cancer/testis antigen 17; CT17; D930038D03Rik; lipase I; lipase member I; lipase, member I; LIPI; LPD lipase; lpd1; LPDL; Membrane-associated phosphatidic acid-selective phospholipase A1-beta; mPA-PLA1 beta; PLA1C; PRED5
Rabbit
Antigen affinity chromatography, Protein A
RUO
149998
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Western Blot
0.45 mg/mL
PBS with 0.09% sodium azide; pH 7.4
Q6XZB0
LIPI
KLH conjugated synthetic peptide between 224-258 amino acids from the Central region of human LIPI.
400 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.