Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lipid A Antibody (26-5), Novus Biologicals™
SDP

Catalog No. NB119881 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB119881 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB119881 Supplier Novus Biologicals Supplier No. NBP311256
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Lipid A Monoclonal specifically detects Lipid A in Bacteria samples. It is validated for Immunoassay.

Specifications

Antigen Lipid A
Applications Immunoassay
Classification Monoclonal
Clone 26-5
Concentration 0.1 mg/ml
Conjugate Unconjugated
Dilution Immunoassay
Formulation 0.2 um filtered solution in PBS, 0.1% BSA
Host Species Mouse
Immunogen The monoclonal antibody 26-5 recognizes lipid A.
Purification Method Protein G purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Bacteria
Content And Storage Store at 4°C. Do not freeze.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.