Learn More
Invitrogen™ Lipid A Monoclonal Antibody (43)

Description
This antibody reacts with the hydrophilic backbone of free lipid A; it only reacts with lipid A not covalently bound to LPS.
Specifications
Specifications
| Antigen | Lipid A |
| Applications | ELISA, Western Blot |
| Classification | Monoclonal |
| Clone | 43 |
| Concentration | 200 μg/mL |
| Conjugate | Unconjugated |
| Formulation | Tissue culture supernatant with 0.02% sodium azide |
| Gene Alias | Lipid A free |
| Host Species | Mouse |
| Immunogen | Lipid A. |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.