Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lipin 1 Antibody - BSA Free, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody has been used in 2 publications

$206.50 - $499.50

Specifications

Antigen Lipin 1
Concentration 0.8 mg/mL
Dilution Western Blot 1.0-2.0 ug/ml, Immunohistochemistry 1:100-1:250, Immunohistochemistry-Paraffin 1:100-1:250
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB11057150T
SDP
View Documents
Novus Biologicals
NB11057150SS
0.025 mL
Each for $206.50
Only null left
Add to Cart
 
NB11057150
SDP
View Documents
Novus Biologicals
NB11057150
0.1 mL
Each for $499.50
Only null left
Add to Cart
 
Description

Description

Lipin 1 Polyclonal specifically detects Lipin 1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

Lipin 1
Western Blot 1.0-2.0 ug/ml, Immunohistochemistry 1:100-1:250, Immunohistochemistry-Paraffin 1:100-1:250
Polyclonal
Rabbit
Cell Biology, Cell Cycle and Replication, Cholesterol Metabolism, Diabetes Research, Lipid and Metabolism, Signal Transduction
PBS and 30% Glycerol with 0.1% Sodium Azide
DKFZp781P1796, EC 3.1.3.18, EC 3.1.3.4, EC 5.4.2.7, KIAA0188phosphatidate phosphatase LPIN1, lipin 1, lipin-1, PAP1
LPIN1
IgG
Affinity Purified
97 kDa
0.8 mg/mL
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Unconjugated
RUO
Human, Mouse, Rat
Q14693
23175
A synthetic peptide to an internal region (within residues 300-400) of the human LPIN1 protein. [Swiss-Prot# Q14693]
Primary
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.