Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lipin 2 Antibody, DyLight 405, Novus Biologicals™
SDP

Catalog No. NB236749 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB236749 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB236749 Supplier Novus Biologicals Supplier No. NBP212783V
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Lipin 2 Polyclonal antibody specifically detects Lipin 2 in Human samples. It is validated for Western Blot

Specifications

Antigen Lipin 2
Applications Western Blot
Classification Polyclonal
Conjugate DyLight 405
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias EC 3.1.3.4, KIAA0249lipin-2, lipin 2, Lipin-2, phosphatidate phosphatase LPIN2
Host Species Rabbit
Immunogen The immunogen this antibody was made to, maps to a region between residue 275 and 325 of human Lipin 2 using the numbering given in entry NP_055461.1 (GeneID 9663).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9663
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.