Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Lipin 2 Antibody, FITC, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP212783F
Description
Lipin 2 Polyclonal antibody specifically detects Lipin 2 in Human samples. It is validated for Western Blot, ImmunoprecipitationSpecifications
Lipin 2 | |
Polyclonal | |
Western Blot, Immunoprecipitation | |
EC 3.1.3.4, KIAA0249lipin-2, lipin 2, Lipin-2, phosphatidate phosphatase LPIN2 | |
The immunogen this antibody was made to, maps to a region between residue 275 and 325 of human Lipin 2 using the numbering given in entry NP_055461.1 (GeneID 9663). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot, Immunoprecipitation | |
FITC | |
PBS | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
9663 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction