Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Lipin 2 Antibody, Novus Biologicals™

Goat Polyclonal Antibody
Supplier: Novus Biologicals NBP300525
Description
Lipin 2 Polyclonal specifically detects Lipin 2 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence, Peptide ELISA.Specifications
Lipin 2 | |
Polyclonal | |
Unconjugated | |
Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA | |
Goat | |
Immunogen affinity purified | |
RUO | |
9663 | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG |
Flow Cytometry, Immunofluorescence, ELISA | |
0.5 mg/mL | |
Flow Cytometry 10 ug/mL, Immunocytochemistry/Immunofluorescence 10 ug/mL, Peptide ELISA Detection limit 1:8000 | |
EC 3.1.3.4, KIAA0249lipin-2, lipin 2, Lipin-2, phosphatidate phosphatase LPIN2 | |
Peptide with sequence CYWRDPIPEVDLDDLS corresponding to C-Terminus according to NP_055461.1. | |
0.1 mg | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction